Learn More
Sino Biological Recombinant Human HIPK4 Protein (Full Length, His Tag), Active
Catalog No. 502574770
Encompass_Preferred
Recombinant full length human HIPK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-4770 Supplier Sino Biological Supplier No. H0610H500
Recombinant full length human HIPK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.