Learn More
Enzo Life Sciences IKK NBD peptide (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP6070500
Encompass_Preferred
The peptide consists of two domains - an antennapedia homeodomain that confers cell permeability and the T735 to E745 region of IKKβ which is the NEMO binding domain (NBD). It is a selective inhibitor of NFκB activation. Blocks interaction of NEMO (an IKB complex regulatory protein) with the IκB kinase complex. The peptide (100 µM) inhibits E-selectin expression in human umbilical vein endothelial cells treated with TNFα and inhibits LPS-induced nitric acid release from a macrophage cell line In an experimental mouse model of acute inflammation, IKK-NBD peptide (topically applied) inhibited PMA induced ear inflammation. Sold under license from Vanderbilt University Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (20mM). Note: This peptide will precipitate when added to tissue culture medium, even to 100µM. However, enough will remain in solution to enter many cell types. Long Term Storage: -20°C.
Catalog No. 50-201-1366