Learn More
Enzo Life Sciences IKK substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2260001
Encompass_Preferred
This peptide, KKKKERLLDDRHDSGLDSMKDEE, was derived from the IκBα sequence. The four lysines on the amino-terminus allow this peptide to be used in phosphocellulose kinase assays for IKK activity. Alternative name: I kappa B kinase substrate. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1295