Learn More
Cayman Chemical IKK2 InhIbItr VI 500ug
SDP
Catalog No. 502980620
Encompass_Preferred
A potent, cell-permeable, reversible inhibitor of IKK2 (IC50 = 13 nM); used to evaluate the role of the canonical, IκB-dependent NF-κB signaling pathway in cellular responses
Catalog No. 50-298-0620 Supplier Cayman Chemical Supplier No. 17276500 UG
A potent, cell-permeable, reversible inhibitor of IKK2 (IC50 = 13 nM); used to evaluate the role of the canonical, IκB-dependent NF-κB signaling pathway in cellular responses