Learn More
Carnabio Usa Inc IKKB(IKBKB)/10UG/TRUNCATED
Catalog No. 507540821
Encompass
IKK(IKBKB), Truncated protein, Wild type, Amino Acid 1-662 , NP_001547.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0821 Supplier Carnabio Usa Inc Supplier No. 0508410
IKK(IKBKB), Truncated protein, Wild type, Amino Acid 1-662 , NP_001547.1, Expressed in Insect (sf21); 10microg