Learn More
Enzo Life Sciences IKKgamma (human) monoclonal antibody (46B844) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804424C100
Encompass_Preferred
Clone: 46B844. Isotype: IgG1. Immunogen: Recombinant human IKKγ (inhibitor of NF-κB kinase γ; IKK3; NEMO). Species reactivity: Human. Applications: WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.2% gelatin and 0.05% sodium azide.
Catalog No. 50-200-9908