Learn More
Cayman Chemical IKKhumnrecombInt 10ug
SDP
Catalog No. 502950025
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged IKKβ expressed in insect cells • Amino acids: 1-756 (full length) • MW: 105 kDa
Catalog No. 50-295-0025 Supplier Cayman Chemical Supplier No. 3373910 UG
Source: Active recombinant human N-terminal GST-tagged IKKβ expressed in insect cells • Amino acids: 1-756 (full length) • MW: 105 kDa