Learn More
Aviva Systems Biology Corp Ikzf1 Antibody - N-terminal region (ARP57849_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57849_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ikzf1. It was validated on Western Blot. . Alternate names: 5832432G11Rik, Ikaros, KIAA4227, LyF-1, Zfpn1a1, Znfn1a1, hlk-1, mKIAA4227ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-403