Learn More
Aviva Systems Biology Corp IKZF1 Antibody - middle region (ARP32758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKZF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IKZF1. Alternate names: Hs.54452, IK1, IKAROS, LYF1, PRO0758, ZNFN1A1, hIk-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54743