Learn More
Aviva Systems Biology Corp IKZF3 Antibody - N-terminal region (ARP39004_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39004_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKZF3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IKZF3. Alternate names: AIO, AIOLOS, ZNFN1A3ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53123