Learn More
Empire Genomics IKZF3 FISH Probe, Red, 20 Tests
Catalog No. NC3725959
Encompass
IKZF3 FISH Probe is designed to detect copy number variations of the IKZF3 gene. The probe can be lableed in any of our standard dyes. Label-Red-dUTP.
Catalog No. NC3725959 Supplier Empire Genomics Supplier No. IKZF320RE
IKZF3 FISH Probe is designed to detect copy number variations of the IKZF3 gene. The probe can be lableed in any of our standard dyes. Label-Red-dUTP.