Learn More
Aviva Systems Biology Corp IKZF4 Antibody - N-terminal region (ARP39553_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39553_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IKZF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human IKZF4. Alternate names: EOS, KIAA1782, ZNFN1A4ApplicationWB, IHC. Species reactivity: Human, Mouse, Pig
Catalog No. 50-014-57083