Learn More
Eurofins Discoverx IL-1α, 2 μg. (92-1153)
Supplier:  Eurofins Discoverx 921153
Encompass
Interleukin 1, alpha (IL-1α) is a pleiotropic cytokine involved in inflammation, immunity, and hematopoiesis. It is a pro-apoptotic protein. Stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. Has pro-inflammatory properties, also promoting fever and sepsis, and has been implicated in multiple diseases related to these symptoms. Interacts with TNF, another pro-inflammatory cytokine. Acts as an agonist for IL1R1. Provided as 2 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9684