Learn More
Medchemexpress LLC IL-1 alpha Human H 5ug
SDP
Catalog No. 5000180591
Encompass_Preferred
IL-1 alpha is a ubiquitous and pivotal pro-inflammatory cytokine IL-1 alpha is produced by monocytes and macrophages as a proprotein which is proteolytically processed and released in response to cell injury and thus induces apoptosis IL-1 alpha plays an important role in inflammation and bridges the innate and adaptive immune systems IL-1 alpha mediates the activation of NF-kappa-B and the three MAPK pathways p38 p42/p44 and JNK pathways[1][4] IL-1 alpha protein Human (HEK293) is a recombinant protein consisting of 159 amino acids (S113-A271) and is produced by HEK293 cells
Catalog No. 50-001-80591 Supplier Medchemexpress LLC Supplier No. HYP731455UG
IL-1 alpha is a ubiquitous and pivotal pro-inflammatory cytokine IL-1 alpha is produced by monocytes and macrophages as a proprotein which is proteolytically processed and released in response to cell injury and thus induces apoptosis IL-1 alpha plays an important role in inflammation and bridges the innate and adaptive immune systems IL-1 alpha mediates the activation of NF-kappa-B and the three MAPK pathways p38 p42/p44 and JNK pathways[1][4] IL-1 alpha protein Human (HEK293) is a recombinant protein consisting of 159 amino acids (S113-A271) and is produced by HEK293 cells