Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-1 alpha ProQuantum Immunoassay Kit
GREENER_CHOICE

Catalog No. p-7161523
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42894 96 Tests
A428945 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A42894 Supplier Invitrogen™ Supplier No. A42894
Only null left
Add to Cart
Add to Cart

ELISA

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01582
Assay Range 0.32 to 5000 pg/mL
Assay Sensitivity <0.3 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene ID (Entrez) 16175
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Storage Requirements Frozen at -20°C
Target IL-1 alpha
Test Time 30 mins
Total Assay Time 2 hrs
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.