Learn More
CPC Scientific 4-(4-Dimethylaminophenylazo)benzoyl-Tyr-Val-Ala-Asp-Ala-Pro-Val-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acid 0.5MG
SDP
Catalog No. 502110492
Encompass
SEQUENCE: 4-(4-Dimethylaminophenylazo)benzoyl-Tyr-Val-Ala-Asp-Ala-Pro-Val-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acidONE-LETTER SEQUENCE: 4-(4-Dimethylaminophenylazo)benzoyl-YVADAPV-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acidMOLECULAR FORMULA: C61H76N12O14S1MOLECULAR WEIGHT: 1233.39STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [161877-70-9]SYNONYMS: Caspase 1 (ICE) Substrate; dabcyl-yvadapv-edans; dabcyl-tyr-val-ala-asp-ala-pro-val-edans; ICE Substrate Ii; il-1-beta Converting Enzyme Substrate Ii; il-1b Converting Enzyme Substrate Ii; 4-(4-dimethylaminophenylazo)benzoyl-tyr-val-ala-asp-ala-pro-val-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acid; 4-(4'-dimethylaminophenylazo)benzoyl-tyrosyl-valyl-alanyl-aspartyl-alanyl-prolyl-valyl-5((2-aminoethyl)amino)naphthalene-1-sulfonic acidRESEARCH AREA: ApoptosisREFERENCES:M.W. Pennington et al., Peptide Res., 7, 72 (1994)M. Los et al., Nature 375, 81 (1995)SKU(s): SUBS-016A, SUBS-016B
Catalog No. 50-211-0492 Supplier CPC Scientific Supplier No. SUBS016A
SEQUENCE: 4-(4-Dimethylaminophenylazo)benzoyl-Tyr-Val-Ala-Asp-Ala-Pro-Val-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acidONE-LETTER SEQUENCE: 4-(4-Dimethylaminophenylazo)benzoyl-YVADAPV-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acidMOLECULAR FORMULA: C61H76N12O14S1MOLECULAR WEIGHT: 1233.39STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [161877-70-9]SYNONYMS: Caspase 1 (ICE) Substrate; dabcyl-yvadapv-edans; dabcyl-tyr-val-ala-asp-ala-pro-val-edans; ICE Substrate Ii; il-1-beta Converting Enzyme Substrate Ii; il-1b Converting Enzyme Substrate Ii; 4-(4-dimethylaminophenylazo)benzoyl-tyr-val-ala-asp-ala-pro-val-5-[(2-aminoethyl)amino]-naphthalene-1-sulfonic acid; 4-(4'-dimethylaminophenylazo)benzoyl-tyrosyl-valyl-alanyl-aspartyl-alanyl-prolyl-valyl-5((2-aminoethyl)amino)naphthalene-1-sulfonic acidRESEARCH AREA: ApoptosisREFERENCES:M.W. Pennington et al., Peptide Res., 7, 72 (1994)M. Los et al., Nature 375, 81 (1995)SKU(s): SUBS-016A, SUBS-016B