Learn More
Medchemexpress LLC IL-1 beta Canine 100ug
SDP
Catalog No. 5000182292
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Canine consists of 152 amino acids (A115-S266) and is expressed in E coli
Catalog No. 50-001-82292 Supplier Medchemexpress LLC Supplier No. HYP73146100UG
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Canine consists of 152 amino acids (A115-S266) and is expressed in E coli