Learn More
Medchemexpress LLC IL-1 beta Canine 10ug
SDP
Catalog No. 5000182208
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Canine consists of 152 amino acids (A115-S266) and is expressed in E coli
Catalog No. 50-001-82208 Supplier Medchemexpress LLC Supplier No. HYP7314610UG
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Canine consists of 152 amino acids (A115-S266) and is expressed in E coli