Learn More
Medchemexpress LLC IL-1 beta Cavia por 10ug
SDP
Catalog No. 5000182554
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Cavia porcellus (His) is produced in E coli with six C-Terminal His-tags It consists of 152 amino acids (T115-S266)
Catalog No. 50-001-82554 Supplier Medchemexpress LLC Supplier No. HYP7257110UG
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Cavia porcellus (His) is produced in E coli with six C-Terminal His-tags It consists of 152 amino acids (T115-S266)