Learn More
Medchemexpress LLC IL-1 beta Cynomolgu 50ug | 50ug
SDP
Catalog No. 5000194003
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Cynomolgus (His) is produced in E coli with a N-Terminal His-tag It consists of 152 amino acids (A117-S268)
Catalog No. 50-001-94003 Supplier Medchemexpress LLC Supplier No. HYP7858550UG
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Cynomolgus (His) is produced in E coli with a N-Terminal His-tag It consists of 152 amino acids (A117-S268)