Learn More
Sigma Aldrich Fine Chemicals Biosciences IL 1 beta human Animal com
Catalog No. 502904748
Encompass
IL-1 (interleukin 1 ) is a proinflammatory cytokine produced in a variety of cells including monocytes tissue macrophages keratinocytes and other epithelial cells. It is synthesized as a pro-IL-1 which is changed to active form IL-1 with the help of pro-inflammatory protease caspase-1. IL-1 belongs to the IL-1 gene family. The gene is mapped to human chromosome 2q14. Recombinant human IL-1 is a 17.3kDa protein containing 153 amino acid residues.
Catalog No. 50-290-4748 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP308310UG
IL-1 (interleukin 1 ) is a proinflammatory cytokine produced in a variety of cells including monocytes tissue macrophages keratinocytes and other epithelial cells. It is synthesized as a pro-IL-1 which is changed to active form IL-1 with the help of pro-inflammatory protease caspase-1. IL-1 belongs to the IL-1 gene family. The gene is mapped to human chromosome 2q14. Recombinant human IL-1 is a 17.3kDa protein containing 153 amino acid residues.