Learn More
Medchemexpress LLC IL-1 beta Human C- 5ug | 5ug
SDP
Catalog No. 5000194390
Encompass_Preferred
IL-1 beta protein is a potent proinflammatory cytokine that plays multiple roles in immune responses It induces inflammatory events including prostaglandin synthesis neutrophil activation and cytokine production IL-1 beta Protein Human (C-His) is the recombinant human-derived IL-1 beta protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-94390 Supplier Medchemexpress LLC Supplier No. HYP78459A5UG
IL-1 beta protein is a potent proinflammatory cytokine that plays multiple roles in immune responses It induces inflammatory events including prostaglandin synthesis neutrophil activation and cytokine production IL-1 beta Protein Human (C-His) is the recombinant human-derived IL-1 beta protein expressed by E coli with C-6 His labeled tag