Learn More
Medchemexpress LLC IL-1 beta Human CH 100ug
SDP
Catalog No. 5000195947
Encompass_Preferred
Recombinant human interleukin-1 beta (IL-1β) produced in CHO cells and supplied lyophilized as a tag-free protein. It is intended for research applications including inflammation and immune signaling studies, functional cell assays, and activity measurements. The material is quality-controlled for purity, endotoxin, and biological potency, and includes formulation and storage recommendations.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) for sensitive assays.
  • Demonstrated bioactivity with ED50 values in the pg/mL range.
  • Lyophilized formulation with stabilizer for improved shelf life.
  • Expressed in mammalian cells to support native-like modifications.
  • Includes sequence and accession information for reproducibility.
  • Recommended aliquoting and storage at -20°C or -80°C for long term.
Catalog No. 50-001-95947 Supplier Medchemexpress LLC Supplier No. HYP7205100UG
Recombinant human interleukin-1 beta (IL-1β) produced in CHO cells and supplied lyophilized as a tag-free protein. It is intended for research applications including inflammation and immune signaling studies, functional cell assays, and activity measurements. The material is quality-controlled for purity, endotoxin, and biological potency, and includes formulation and storage recommendations.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) for sensitive assays.
  • Demonstrated bioactivity with ED50 values in the pg/mL range.
  • Lyophilized formulation with stabilizer for improved shelf life.
  • Expressed in mammalian cells to support native-like modifications.
  • Includes sequence and accession information for reproducibility.
  • Recommended aliquoting and storage at -20°C or -80°C for long term.