Learn More
Medchemexpress LLC IL-1 beta Human E6 10ug
SDP
Catalog No. 5000182491
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Human (E6K His) is produced in E coli with a N-Terminal His-tag It consists of 269 amino acids (M1-S269)
Catalog No. 50-001-82491 Supplier Medchemexpress LLC Supplier No. HYP7314910UG
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Human (E6K His) is produced in E coli with a N-Terminal His-tag It consists of 269 amino acids (M1-S269)