Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-1 beta protein, human (N-His) | 96.9% | 10UG
SDP

Catalog No. 5000177791
Encompass_Preferred

Recombinant human interleukin-1 beta (IL-1β) with an N-terminal 6xHis tag, supplied as a purified protein for in vitro research. The protein is expressed in E. coli and characterized by SDS-PAGE, and is intended for studies of inflammation, cytokine signaling, and cell activation.

  • Recombinant human origin with N-terminal 6xHis tag for purification and detection.
  • High purity: 96.9% (reported).
  • Predicted molecular mass 17.4 kDa (≈18 kDa by SDS-PAGE).
  • Expressed in E. coli expression system.
  • Supplied as a small-scale research quantity suitable for biochemical and cell-based assays.
  • Store at -20°C to maintain stability.

Catalog No. 50-001-77791 Supplier Medchemexpress LLC Supplier No. HYP70025410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-1 beta (IL-1β) with an N-terminal 6xHis tag, supplied as a purified protein for in vitro research. The protein is expressed in E. coli and characterized by SDS-PAGE, and is intended for studies of inflammation, cytokine signaling, and cell activation.

  • Recombinant human origin with N-terminal 6xHis tag for purification and detection.
  • High purity: 96.9% (reported).
  • Predicted molecular mass 17.4 kDa (≈18 kDa by SDS-PAGE).
  • Expressed in E. coli expression system.
  • Supplied as a small-scale research quantity suitable for biochemical and cell-based assays.
  • Store at -20°C to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.