Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-1 beta (recombinant human, N-His tag) | 2UG
SDP

Catalog No. 5000177420
Encompass_Preferred

Recombinant human interleukin-1 beta (IL-1β) with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder. Intended for cell-based assays and biochemical studies, the protein is characterized for low endotoxin and defined biological activity, and is formulated for stability and long-term storage.

  • Recombinant human IL-1β encompassing residues A117-S269.
  • N-terminal His tag for purification and detection.
  • Expressed in Escherichia coli and provided as lyophilized powder.
  • Approx. 18 kDa molecular weight by SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • High specific activity (ED50 1.045 pg/mL in CTLL-2 assay).
  • Formulated in 50 mM Tris-HCl, 300 mM NaCl, 200 mM arginine, pH 8.0.
  • Store lyophilized at -20°C; reconstitute ≥100 μg/mL and add carrier for long-term storage.

Catalog No. 50-001-77420 Supplier Medchemexpress LLC Supplier No. HYP7002542UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-1 beta (IL-1β) with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder. Intended for cell-based assays and biochemical studies, the protein is characterized for low endotoxin and defined biological activity, and is formulated for stability and long-term storage.

  • Recombinant human IL-1β encompassing residues A117-S269.
  • N-terminal His tag for purification and detection.
  • Expressed in Escherichia coli and provided as lyophilized powder.
  • Approx. 18 kDa molecular weight by SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • High specific activity (ED50 1.045 pg/mL in CTLL-2 assay).
  • Formulated in 50 mM Tris-HCl, 300 mM NaCl, 200 mM arginine, pH 8.0.
  • Store lyophilized at -20°C; reconstitute ≥100 μg/mL and add carrier for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.