Learn More
Medchemexpress LLC Interleukin-1 beta protein, mouse (His, Avi) | >95.0% | 20UG
SDP
Catalog No. 5000176129
Encompass_Preferred
Recombinant mouse interleukin-1 beta (IL-1β) with N-terminal Avi and 8xHis tags, expressed in E. coli and supplied lyophilized. The protein corresponds to residues V118-S269 and is formulated for stability and use in immunoassays, receptor binding studies, and bioactivity assays.
  • N-terminal Avi and 8xHis tags for affinity purification and detection.
  • Expressed in E. coli to provide a bacterial-derived recombinant protein.
  • High purity (>95%) as determined by Tris-Bis PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Apparent molecular weight ~21.0 kDa by SDS-PAGE.
  • Recommended storage at -20 °C to -80 °C; avoid repeated freeze-thaw cycles.
  • Suitable for immunoassays, binding studies, and functional bioactivity assays.
Catalog No. 50-001-76129 Supplier Medchemexpress LLC Supplier No. HYP70396920UG
Recombinant mouse interleukin-1 beta (IL-1β) with N-terminal Avi and 8xHis tags, expressed in E. coli and supplied lyophilized. The protein corresponds to residues V118-S269 and is formulated for stability and use in immunoassays, receptor binding studies, and bioactivity assays.
  • N-terminal Avi and 8xHis tags for affinity purification and detection.
  • Expressed in E. coli to provide a bacterial-derived recombinant protein.
  • High purity (>95%) as determined by Tris-Bis PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Apparent molecular weight ~21.0 kDa by SDS-PAGE.
  • Recommended storage at -20 °C to -80 °C; avoid repeated freeze-thaw cycles.
  • Suitable for immunoassays, binding studies, and functional bioactivity assays.