Learn More
Medchemexpress LLC Recombinant mouse interleukin-1 beta (His, Avi) | >95.0% | 50UG
SDP
Catalog No. 5000174994
Encompass_Preferred
Recombinant mouse interleukin-1 beta (mature form V118-S269) is a potent proinflammatory cytokine produced in Escherichia coli and supplied lyophilized with N-terminal Avi and 8xHis tags. It is intended for in vitro immune signaling studies, cytokine assays, and receptor activation experiments, and is characterized by high purity and low endotoxin to support sensitive biological assays.
  • High purity >95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg (LAL method).
  • Expressed in Escherichia coli and corresponds to residues V118-S269.
  • Supplied lyophilized with N-terminal Avi and 8xHis tags.
  • Recommended storage: lyophilized at -20°C; aliquot and store at -80°C after reconstitution.
Catalog No. 50-001-74994 Supplier Medchemexpress LLC Supplier No. HYP70396950UG
Recombinant mouse interleukin-1 beta (mature form V118-S269) is a potent proinflammatory cytokine produced in Escherichia coli and supplied lyophilized with N-terminal Avi and 8xHis tags. It is intended for in vitro immune signaling studies, cytokine assays, and receptor activation experiments, and is characterized by high purity and low endotoxin to support sensitive biological assays.
  • High purity >95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg (LAL method).
  • Expressed in Escherichia coli and corresponds to residues V118-S269.
  • Supplied lyophilized with N-terminal Avi and 8xHis tags.
  • Recommended storage: lyophilized at -20°C; aliquot and store at -80°C after reconstitution.