Learn More
Medchemexpress LLC IL-1 beta Pig 100ug
SDP
Catalog No. 5000196303
Encompass_Preferred
Recombinant porcine interleukin-1 beta (IL-1β) supplied as a tag-free protein produced in E. coli for in vitro research. Supplied quantity is 100 μg and the product is characterized by high purity (≥95% by reducing SDS-PAGE). Certificate of analysis recommends reconstituting to at least 100 μg/mL in ddH2O. Intended for use in studies of inflammation, cytokine activity, and immune signaling.
  • High purity (≥95%) ensures minimal contaminants.
  • Tag free to reduce assay interference.
  • Produced in E. coli for recombinant expression consistency.
  • Supplied as 100 μg suitable for small-scale experiments.
  • Recommended reconstitution at or above 100 μg/mL to preserve handling stability.
Catalog No. 50-001-96303 Supplier Medchemexpress LLC Supplier No. HYP7206100UG
Recombinant porcine interleukin-1 beta (IL-1β) supplied as a tag-free protein produced in E. coli for in vitro research. Supplied quantity is 100 μg and the product is characterized by high purity (≥95% by reducing SDS-PAGE). Certificate of analysis recommends reconstituting to at least 100 μg/mL in ddH2O. Intended for use in studies of inflammation, cytokine activity, and immune signaling.
  • High purity (≥95%) ensures minimal contaminants.
  • Tag free to reduce assay interference.
  • Produced in E. coli for recombinant expression consistency.
  • Supplied as 100 μg suitable for small-scale experiments.
  • Recommended reconstitution at or above 100 μg/mL to preserve handling stability.