Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-1 beta Pig 2ug
SDP

Catalog No. 5000195966
Encompass_Preferred

Recombinant porcine interleukin-1 beta (IL-1β) is a proinflammatory cytokine provided as a tag-free, E. coli-derived protein for use in inflammation and immunology research. The product is characterized for purity and molecular weight, and includes batch-specific quality documentation and storage guidance suitable for assay preparation and validation.

  • Tag-free recombinant protein for minimal interference in assays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 17.88 kDa for identification and QC.
  • Batch-specific certificate of analysis included for traceable quality.
  • Storage and handling guidance to preserve activity, including aliquoting and carrier protein recommendations for long-term storage.

Catalog No. 50-001-95966 Supplier Medchemexpress LLC Supplier No. HYP72062UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant porcine interleukin-1 beta (IL-1β) is a proinflammatory cytokine provided as a tag-free, E. coli-derived protein for use in inflammation and immunology research. The product is characterized for purity and molecular weight, and includes batch-specific quality documentation and storage guidance suitable for assay preparation and validation.

  • Tag-free recombinant protein for minimal interference in assays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 17.88 kDa for identification and QC.
  • Batch-specific certificate of analysis included for traceable quality.
  • Storage and handling guidance to preserve activity, including aliquoting and carrier protein recommendations for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.