Learn More
Medchemexpress LLC IL-1 beta Rat 100ug
SDP
Catalog No. 5000197174
Encompass_Preferred
Recombinant rat interleukin-1 beta supplied as a lyophilized powder for research use in inflammation and immunology. The protein is expressed in E. coli, provided with characterized purity and low endotoxin, and intended for in vitro assays and protein characterization after reconstitution following the manufacturer's datasheet.
  • Recombinant rat interleukin-1 beta for inflammation research.
  • High purity greater than 95% for reliable assay performance.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive bioassays.
  • Lyophilized powder for extended storage stability.
  • Expression in E. coli with accession-defined sequence for reproducibility.
Catalog No. 50-001-97174 Supplier Medchemexpress LLC Supplier No. HYP7097100UG
Recombinant rat interleukin-1 beta supplied as a lyophilized powder for research use in inflammation and immunology. The protein is expressed in E. coli, provided with characterized purity and low endotoxin, and intended for in vitro assays and protein characterization after reconstitution following the manufacturer's datasheet.
  • Recombinant rat interleukin-1 beta for inflammation research.
  • High purity greater than 95% for reliable assay performance.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive bioassays.
  • Lyophilized powder for extended storage stability.
  • Expression in E. coli with accession-defined sequence for reproducibility.