Learn More
Medchemexpress LLC IL-1 beta Rhesus ma 100ug
SDP
Catalog No. 5000195543
Encompass_Preferred
Recombinant interleukin-1 beta (IL-1β) from rhesus macaque supplied as a purified protein for in vitro immunology and cell signaling research. The product contains the mature cytokine sequence (153 amino acids, A117-S269), is reported at ≥98% purity, and has an approximate molecular weight of 18.4 kDa by SDS-PAGE. Intended for research use to study inflammatory signaling and cytokine responses.
  • Contains mature cytokine sequence (153 amino acids, A117-S269).
  • High purity (≥98%) reduces background in downstream assays.
  • Approximate molecular weight ~18.4 kDa confirmed by SDS-PAGE.
  • Suitable for in vitro immunology and cell signaling studies.
  • Available in small-scale quantities for assay development (100 μg).
Catalog No. 50-001-95543 Supplier Medchemexpress LLC Supplier No. HYP7207100UG
Recombinant interleukin-1 beta (IL-1β) from rhesus macaque supplied as a purified protein for in vitro immunology and cell signaling research. The product contains the mature cytokine sequence (153 amino acids, A117-S269), is reported at ≥98% purity, and has an approximate molecular weight of 18.4 kDa by SDS-PAGE. Intended for research use to study inflammatory signaling and cytokine responses.
  • Contains mature cytokine sequence (153 amino acids, A117-S269).
  • High purity (≥98%) reduces background in downstream assays.
  • Approximate molecular weight ~18.4 kDa confirmed by SDS-PAGE.
  • Suitable for in vitro immunology and cell signaling studies.
  • Available in small-scale quantities for assay development (100 μg).