Learn More
Medchemexpress LLC IL-1 beta Rhesus ma 20ug
SDP
Catalog No. 5000195882
Encompass_Preferred
Recombinant rhesus macaque interleukin-1 beta (IL-1β) protein, representing the mature cytokine sequence and supplied as a purified recombinant preparation for in vitro immunology and inflammation research. The product is expressed in E. coli, characterized at roughly 153 amino acids (mature residues A117-S269), and is commonly provided in small pack sizes for assay development and biochemical studies.
  • Recombinant mature cytokine sequence A117-S269, ~153 amino acids.
  • Expressed in E. coli for efficient production.
  • High purity suitable for in vitro assays and biochemical characterization.
  • Approximate molecular weight ~18.4 kDa by SDS-PAGE.
  • Available in small pack sizes for assay development and pilot studies.
Catalog No. 50-001-95882 Supplier Medchemexpress LLC Supplier No. HYP720720UG
Recombinant rhesus macaque interleukin-1 beta (IL-1β) protein, representing the mature cytokine sequence and supplied as a purified recombinant preparation for in vitro immunology and inflammation research. The product is expressed in E. coli, characterized at roughly 153 amino acids (mature residues A117-S269), and is commonly provided in small pack sizes for assay development and biochemical studies.
  • Recombinant mature cytokine sequence A117-S269, ~153 amino acids.
  • Expressed in E. coli for efficient production.
  • High purity suitable for in vitro assays and biochemical characterization.
  • Approximate molecular weight ~18.4 kDa by SDS-PAGE.
  • Available in small pack sizes for assay development and pilot studies.