Learn More
Medchemexpress LLC IL-1 beta Rhesus ma 500ug
SDP
Catalog No. 5000195556
Encompass_Preferred
Recombinant rhesus macaque interleukin-1 beta (IL-1β) representing the mature cytokine sequence (A117-S269; 153 amino acids). The protein is expressed in Escherichia coli and supplied for research use in studies of inflammation, immune signaling, and cytokine biology.
  • High purity for research applications.
  • Corresponds to the mature rhesus IL-1β sequence (A117-S269).
  • Expressed in E. coli for recombinant production.
  • Suitable for cytokine assays and signaling studies.
  • Supplied in research-scale quantities (e.g., 500 μg).
Catalog No. 50-001-95556 Supplier Medchemexpress LLC Supplier No. HYP7207500UG
Recombinant rhesus macaque interleukin-1 beta (IL-1β) representing the mature cytokine sequence (A117-S269; 153 amino acids). The protein is expressed in Escherichia coli and supplied for research use in studies of inflammation, immune signaling, and cytokine biology.
  • High purity for research applications.
  • Corresponds to the mature rhesus IL-1β sequence (A117-S269).
  • Expressed in E. coli for recombinant production.
  • Suitable for cytokine assays and signaling studies.
  • Supplied in research-scale quantities (e.g., 500 μg).