Learn More
Medchemexpress LLC HY-P7074A 10ug Medchemexpress, IL-10 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934317
Encompass_Preferred
Medchemexpress, HY-P7074A 10ug IL-10 (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-10 (CHO-expressed) is a potent CHO cell derived anti-inflammatory cytokine results in increased immune-mediated demyelination in mice infected with a neurotropic coronavirus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4317 Supplier Medchemexpress LLC Supplier No. HYP7074A10UG
Medchemexpress, HY-P7074A 10ug IL-10 (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-10 (CHO-expressed) is a potent CHO cell derived anti-inflammatory cytokine results in increased immune-mediated demyelination in mice infected with a neurotropic coronavirus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.