Learn More
Enzo Life Sciences IL-10 (human), ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901036
Encompass_Preferred
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce laboratory errors. Alternative name: Interleukin-10. Sensitivity: 3.75 pg/ml (range 7.81 - 500 pg/ml). Use/Stability: Store all components at +4°C, except standard at -20°C. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6889