Learn More
Enzo Life Sciences IL-10 (human), (recombinant) (10µg)
SDP
Catalog No. 502013926
Encompass_Preferred
Alternative name: Interleukin-10, Cytokine synthesis inhibitory factor, CSIF. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 5mM sodium phosphate, pH 7.2. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3926 Supplier Enzo Life Sciences Supplier No. ENZPRT1340010
Alternative name: Interleukin-10, Cytokine synthesis inhibitory factor, CSIF. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 5mM sodium phosphate, pH 7.2. Source: Produced in E.coli. Long Term Storage: -20°C.