Learn More
Enzo Life Sciences IL-10 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900036
Encompass_Preferred
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce laboratory errors. Alternative Name: Interleukin-10. Sensitivity: 3.75 pg/ml (range 7.81 - 500 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution
Catalog No. 50-658-113