Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-10 Human HEK293 100ug
SDP

Catalog No. 5000197063
Encompass_Preferred

Recombinant human interleukin-10 produced in HEK293 cells, supplied as a lyophilized, glycosylated protein for use in immunology and cell signaling research. Supplied as a 100 μg vial with low endotoxin and high purity; recommended for binding and activity assays, cell culture supplementation, and assay development.

  • Glycosylated human IL-10 expressed in HEK293 cells.
  • Supplied as lyophilized powder for long-term storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL).
  • Predicted mass ~18.6 kDa; observed ~16-20 kDa on SDS-PAGE.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Stable at -20°C for up to 2 years; aliquot for long-term storage.
  • Suitable for binding and activity assays (reported EC50 0.3113 μg/mL).

Catalog No. 50-001-97063 Supplier Medchemexpress LLC Supplier No. HYP70751100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-10 produced in HEK293 cells, supplied as a lyophilized, glycosylated protein for use in immunology and cell signaling research. Supplied as a 100 μg vial with low endotoxin and high purity; recommended for binding and activity assays, cell culture supplementation, and assay development.

  • Glycosylated human IL-10 expressed in HEK293 cells.
  • Supplied as lyophilized powder for long-term storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL).
  • Predicted mass ~18.6 kDa; observed ~16-20 kDa on SDS-PAGE.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Stable at -20°C for up to 2 years; aliquot for long-term storage.
  • Suitable for binding and activity assays (reported EC50 0.3113 μg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.