Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-10 Human HEK293 10ug
SDP

Catalog No. 5000184414
Encompass_Preferred

Recombinant human interleukin-10 (IL-10) produced in HEK293 cells and supplied lyophilized as a tag-free protein for research use. The preparation is >95% pure by reducing SDS-PAGE, with a predicted molecular mass of 18.6 kDa and is lyophilized from PBS or 20 mM PB, 150 mM NaCl. Intended for receptor-binding and functional immunology studies; aliquot and store at -20°C or -80°C for extended stability.

  • Expression system suitable for human-like post-translational modifications.
  • Tag-free format to avoid interference in binding studies.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for convenient storage and transport.
  • Predicted molecular mass ~18.6 kDa with glycosylation-dependent mobility.
  • Suitable for receptor binding and functional assays; EC50 reported by manufacturer.

Catalog No. 50-001-84414 Supplier Medchemexpress LLC Supplier No. HYP7075110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-10 (IL-10) produced in HEK293 cells and supplied lyophilized as a tag-free protein for research use. The preparation is >95% pure by reducing SDS-PAGE, with a predicted molecular mass of 18.6 kDa and is lyophilized from PBS or 20 mM PB, 150 mM NaCl. Intended for receptor-binding and functional immunology studies; aliquot and store at -20°C or -80°C for extended stability.

  • Expression system suitable for human-like post-translational modifications.
  • Tag-free format to avoid interference in binding studies.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for convenient storage and transport.
  • Predicted molecular mass ~18.6 kDa with glycosylation-dependent mobility.
  • Suitable for receptor binding and functional assays; EC50 reported by manufacturer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.