Learn More
Medchemexpress LLC IL-10 Human HEK293 10ug
SDP
Supplier:  Medchemexpress LLC HYP7315310UG
Encompass_Preferred
IL-10 protein is a major immunoregulatory cytokine with profound anti-inflammatory functions that limits excessive tissue destruction caused by inflammation Mechanistically IL10 binds to its heterotetrameric receptors IL10RA and IL10RB leading to JAK1- and STAT2-mediated phosphorylation of STAT3 which translocates to the nucleus and drives the expression of anti-inflammatory mediators IL-10 Protein Human (HEK293 His) is the recombinant human-derived IL-10 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-82199