Learn More
AdipoGen IL-10 (mouse):Fc (mouse) (rec.) (non-lytic)
SDP
Supplier:  AdipoGen CHIMF12010C050
Encompass_Preferred
IL-10 (mouse):Fc (mouse) (rec.) (non-lytic), AdipoGen Life Sciences. Protein. The extracellular domain of mouse IL-10 (aa 1-178) is fused to the N-terminus of the Fc region of a mutant mouse IgG2a. Shows the biological function of the IL-10 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain. Lyophilized from 0.2¿m-filtered solution in PBS.
Catalog No. 50-117-6288