Learn More
Medchemexpress LLC IL-10 Rat CHO 100ug
SDP
Catalog No. 5000196844
Encompass_Preferred
IL-10 Protein, Rat (CHO) is a CHO cell-derived recombinant rat interleukin-10 cytokine supplied as a tag-free, glycosylated protein for in vitro research. It is intended for studies of anti-inflammatory and immunomodulatory responses and is quality controlled for purity, endotoxin, and bioactivity.
  • CHO-expressed, tag free glycosylated protein suitable for cell-based assays.
  • High purity (≥95%) verified by reducing SDS-PAGE.
  • Low endotoxin (<0.2 EU/μg) for sensitive biological assays.
  • Bioactivity validated with ED50 <8 μg/mL in a C6 cell assay.
  • Stable when stored frozen; add carrier protein for long-term storage.
Catalog No. 50-001-96844 Supplier Medchemexpress LLC Supplier No. HYP7098A100UG
IL-10 Protein, Rat (CHO) is a CHO cell-derived recombinant rat interleukin-10 cytokine supplied as a tag-free, glycosylated protein for in vitro research. It is intended for studies of anti-inflammatory and immunomodulatory responses and is quality controlled for purity, endotoxin, and bioactivity.
  • CHO-expressed, tag free glycosylated protein suitable for cell-based assays.
  • High purity (≥95%) verified by reducing SDS-PAGE.
  • Low endotoxin (<0.2 EU/μg) for sensitive biological assays.
  • Bioactivity validated with ED50 <8 μg/mL in a C6 cell assay.
  • Stable when stored frozen; add carrier protein for long-term storage.