Learn More
Medchemexpress LLC IL-10 Rat CHO 10ug
SDP
Catalog No. 5000196845
Encompass_Preferred
Recombinant rat interleukin-10 (IL-10), produced in CHO cells and supplied lyophilized (10 μg), is an immunosuppressive cytokine used to investigate anti-inflammatory signaling, immune regulation, and cytokine-mediated cellular responses. It is intended for in vitro applications such as cell stimulation, signaling pathway analysis, and functional assays; reconstitute and store aliquots per datasheet recommendations.
  • CHO cell expression preserves eukaryotic post-translational modifications.
  • Lyophilized for long-term stability and convenient reconstitution.
  • Suitable for cell culture stimulation and functional assays.
  • Provided with quality documentation including certificate of analysis.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.
Catalog No. 50-001-96845 Supplier Medchemexpress LLC Supplier No. HYP7098A10UG
Recombinant rat interleukin-10 (IL-10), produced in CHO cells and supplied lyophilized (10 μg), is an immunosuppressive cytokine used to investigate anti-inflammatory signaling, immune regulation, and cytokine-mediated cellular responses. It is intended for in vitro applications such as cell stimulation, signaling pathway analysis, and functional assays; reconstitute and store aliquots per datasheet recommendations.
  • CHO cell expression preserves eukaryotic post-translational modifications.
  • Lyophilized for long-term stability and convenient reconstitution.
  • Suitable for cell culture stimulation and functional assays.
  • Provided with quality documentation including certificate of analysis.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.