Learn More
Medchemexpress LLC IL-10R beta Human 50ug
SDP
Catalog No. 5000181370
Encompass_Preferred
IL-10R beta protein is an IL10 cytokine surface receptor involved in IL10-mediated inflammation and immune regulation as well as viral defense responses IL-10R beta Protein Human (HEK293 His) is expressed by HEK 293 cells and has a transmembrane region (W221-F242) with a 6 His tag at the C-terminus[3]
Catalog No. 50-001-81370 Supplier Medchemexpress LLC Supplier No. HYP7259950UG
IL-10R beta protein is an IL10 cytokine surface receptor involved in IL10-mediated inflammation and immune regulation as well as viral defense responses IL-10R beta Protein Human (HEK293 His) is expressed by HEK 293 cells and has a transmembrane region (W221-F242) with a 6 His tag at the C-terminus[3]