Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-10R beta protein, rat (HEK293, Fc) | >95.0% | 10 UG
SDP

Catalog No. 5000290381
Encompass_Preferred

Recombinant rat interleukin-10 receptor subunit beta extracellular domain, expressed in HEK293 cells and fused to a human Fc tag. Supplied lyophilized for use in immunoassays, binding studies, and receptor interaction experiments. Formulated for low endotoxin and high purity to support sensitive assays.

  • Expressed in HEK293 cells with a C-terminal human Fc tag.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Approximate molecular weight 68 kDa.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen aliquots for long-term storage.

Catalog No. 50-002-90381 Supplier Medchemexpress LLC Supplier No. HYP7641410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-10 receptor subunit beta extracellular domain, expressed in HEK293 cells and fused to a human Fc tag. Supplied lyophilized for use in immunoassays, binding studies, and receptor interaction experiments. Formulated for low endotoxin and high purity to support sensitive assays.

  • Expressed in HEK293 cells with a C-terminal human Fc tag.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Approximate molecular weight 68 kDa.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen aliquots for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.