Learn More
Medchemexpress LLC Il-10r beta protein, rat (HEK293, Fc) | >95.0% | 2 UG
SDP
Catalog No. 5000290007
Encompass_Preferred
Recombinant rat interleukin-10 receptor subunit beta produced in HEK293 mammalian cells with a C-terminal human Fc tag, supplied as a purified protein for research use. The product is provided as a 2 μg vial with >95% purity and is suitable for receptor binding, signaling assays, and other in vitro studies.
  • Expressed in HEK293 mammalian cells to support native folding.
  • C-terminal human Fc tag facilitates detection and purification.
  • >95.0% purity as determined by reducing SDS-PAGE.
  • Suitable for receptor binding and signaling assays, and biochemical characterization.
  • Store at -20°C; aliquot and avoid repeated freeze-thaw cycles.
Catalog No. 50-002-90007 Supplier Medchemexpress LLC Supplier No. HYP764142UG
Recombinant rat interleukin-10 receptor subunit beta produced in HEK293 mammalian cells with a C-terminal human Fc tag, supplied as a purified protein for research use. The product is provided as a 2 μg vial with >95% purity and is suitable for receptor binding, signaling assays, and other in vitro studies.
  • Expressed in HEK293 mammalian cells to support native folding.
  • C-terminal human Fc tag facilitates detection and purification.
  • >95.0% purity as determined by reducing SDS-PAGE.
  • Suitable for receptor binding and signaling assays, and biochemical characterization.
  • Store at -20°C; aliquot and avoid repeated freeze-thaw cycles.