Learn More
Biotium IL-10R1, Mouse (Interleukin-10 Receptor 1) (CD210)(1B1.3a), CF568 conjugate, 0.1mg/mL
Supplier:  Biotium BNC682001500
Encompass_Preferred
This monoclonal antibody recognizes Mouse Interleukin-10 receptor 1 (IL-10R1), a cell surface glycoprotein of approximately 110 kDa. Its epitope is localized in the ligand-binding domain in the extracellular region of Mouse IL-10R1. This antibody blocks the binding of IL-10 to its receptor. IL-10R1 is a class II cytokine receptor expressed on thymocytes, T cells, B cells, NK cells, monocytes and macrophages. Binding of IL-10 to its receptor initiates JAK/STAT signaling that leads to B cell proliferation and inhibition of macrophage activation. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-006-46744