Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-10RB ELISA Kit

Catalog No. EHIL10RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHIL10RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHIL10RBX10 Supplier Invitrogen™ Supplier No. EHIL10RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The Human Interleukin 10 Receptor Beta (Hu IL-10RB) ELISA quantitates Hu IL-10RB in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-10RB. Principle of the method The Human IL-10RB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08334
Assay Range 5-3000 pg/mL
Assay Sensitivity 5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD15603736620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2
Gene ID (Entrez) 3588
Gene Symbol IL10RB
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL10RB
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.