Learn More
Medchemexpress LLC HY-P7031A 10ug Medchemexpress, IL-11 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083096
Encompass_Preferred
Medchemexpress, HY-P7031A 10ug IL-11 (CHO-expressed), Human CAS: Recombinant Human Interleukin-11 (CHO-expressed) is a CHO cell derived protective factor, which has a protective role and can accelerate recovery of platelets, and remarkably lessen the extent of inflammatory responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3096 Supplier Medchemexpress LLC Supplier No. HYP7031A10UG
Medchemexpress, HY-P7031A 10ug IL-11 (CHO-expressed), Human CAS: Recombinant Human Interleukin-11 (CHO-expressed) is a CHO cell derived protective factor, which has a protective role and can accelerate recovery of platelets, and remarkably lessen the extent of inflammatory responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.